siRNA Details
| Gene Symbol: LOC105371899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136416.1 | 3 | 661 | |||
| GenBank mRNA | BM700476.1 | - | 3 | 559 | ||
| CD675441.1 | - | 3 | 502 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371899| Gene Symbol: LOC105371899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136416.1 | 3 | 661 | |||
| GenBank mRNA | BM700476.1 | - | 3 | 559 | ||
| CD675441.1 | - | 3 | 502 | |||