siRNA Details
| Gene Symbol: LOC101927557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110808.1 | 3 | 496 | |||
| GenBank mRNA | AK131022.1 | - | 3 | 496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927557| Gene Symbol: LOC101927557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110808.1 | 3 | 496 | |||
| GenBank mRNA | AK131022.1 | - | 3 | 496 | ||