siRNA Details
| Gene Symbol: LOC102546294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105057.1 | 5 | 1425 | |||
| GenBank mRNA | AK054753.1 | - | 5 | 1358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102546294| Gene Symbol: LOC102546294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105057.1 | 5 | 1425 | |||
| GenBank mRNA | AK054753.1 | - | 5 | 1358 | ||