siRNA Details
Gene Symbol: LINC01619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046159.2 | 6 | 1325 | |||
NR_046160.2 | 6 | 1487 | ||||
NR_132339.1 | 5 | 1563 | ||||
NR_132340.1 | 5 | 1412 | ||||
NR_132341.1 | 5 | 1435 | ||||
GenBank mRNA | AK055439.1 | - | 5 | 1403 | ||
BC029844.1 | - | 6 | 1325 | |||
BC107595.1 | - | 5 | 1249 | |||
BC118985.1 | - | 6 | 1323 | |||
BC122534.1 | - | 5 | 1272 |
