siRNA Details
| Gene Symbol: SMG7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040063.1 | 5 | 2020 | |||
| GenBank mRNA | BC032873.2 | - | 5 | 1784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMG7-AS1| Gene Symbol: SMG7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040063.1 | 5 | 2020 | |||
| GenBank mRNA | BC032873.2 | - | 5 | 1784 | ||