siRNA Details
| Gene Symbol: LOC102723640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135684.1 | 4 | 590 | |||
| NR_135685.1 | 3 | 472 | ||||
| GenBank mRNA | BG203382.1 | - | 1 | 237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX096897.1 | - | 2 | 334 | Chr. 15 - 72608486 - 72616230 [-] on Build GRCh38 | |

LOC102723640
Genomic Map