siRNA Details
| Gene Symbol: KCCAT333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110014.1 | 2 | 188 | |||
| GenBank mRNA | BX114378.1 | - | 2 | 188 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG287431.1 | - | 242 | Chr. - - [-] on Build | ||

KCCAT333
Genomic Map