siRNA Details
| Gene Symbol: LOC101929741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125930.1 | 3 | 122 | |||
| GenBank mRNA | BF507886.1 | - | 3 | 109 | ||
| BQ438340.1 | - | 3 | 123 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM546510.1 | - | 130 | Chr. - - [-] on Build | ||

LOC101929741
Genomic Map