siRNA Details
| Gene Symbol: SH3PXD2A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038940.1 | 3 | 832 | |||
| GenBank mRNA | AK056784.1 | - | 3 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3PXD2A-AS1| Gene Symbol: SH3PXD2A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038940.1 | 3 | 832 | |||
| GenBank mRNA | AK056784.1 | - | 3 | 614 | ||