siRNA Details
Gene Symbol: SYNE1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120501.1 | 1 | 50 | |||
GenBank mRNA | CK821189.1 | - | 1 | 50 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SYNE1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120501.1 | 1 | 50 | |||
GenBank mRNA | CK821189.1 | - | 1 | 50 |