siRNA Details
| Gene Symbol: SYNE1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120501.1 | 2 | 492 | |||
| GenBank mRNA | CK821189.1 | - | 2 | 492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYNE1-AS1| Gene Symbol: SYNE1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120501.1 | 2 | 492 | |||
| GenBank mRNA | CK821189.1 | - | 2 | 492 | ||