siRNA Details
| Gene Symbol: LOC102724714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135567.1 | 2 | 140 | |||
| GenBank mRNA | AW969319.1 | - | 2 | 140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724714| Gene Symbol: LOC102724714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135567.1 | 2 | 140 | |||
| GenBank mRNA | AW969319.1 | - | 2 | 140 | ||