siRNA Details
| Gene Symbol: LINC01280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110392.1 | 4 | 1118 | |||
| GenBank mRNA | AK307257.1 | - | 4 | 1118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01280| Gene Symbol: LINC01280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110392.1 | 4 | 1118 | |||
| GenBank mRNA | AK307257.1 | - | 4 | 1118 | ||