siRNA Details
Gene Symbol: LOC728175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040108.1 | 1 | 322 | |||
GenBank mRNA | AK097110.1 | - | 1 | 322 | ||
BC020886.1 | - | 1 | 221 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC728175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040108.1 | 1 | 322 | |||
GenBank mRNA | AK097110.1 | - | 1 | 322 | ||
BC020886.1 | - | 1 | 221 |