siRNA Details
| Gene Symbol: LOC148413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015434.1 | 3 | 1649 | |||
| GenBank mRNA | AK024040.1 | - | 3 | 1623 | ||
| AK092572.1 | - | 1 | 2142 | |||
| BC068600.1 | - | 3 | 1606 | |||
| BC080546.1 | - | 3 | 1605 | |||
| Gene Symbol: MRPL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA824533.1 | - | 1 | 57 | ||
| AW015403.1 | - | 1 | 68 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY250895.1 | - | 1 | 70 | Chr. 1 - 1401909 - 1406964 [+] on Build GRCh38 | |

LOC148413
Genomic Map