siRNA Details
| Gene Symbol: NDUFV2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110771.1 | 3 | 521 | |||
| GenBank mRNA | BI520375.1 | - | 3 | 446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY038625.1 | - | 3 | 440 | Chr. 18 - 9132328 - 9136639 [-] on Build GRCh38 | |

NDUFV2-AS1
Genomic Map