siRNA Details
| Gene Symbol: LOC101927901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110063.1 | 5 | 2012 | |||
| GenBank mRNA | BC035381.1 | - | 5 | 1978 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927901| Gene Symbol: LOC101927901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110063.1 | 5 | 2012 | |||
| GenBank mRNA | BC035381.1 | - | 5 | 1978 | ||