siRNA Details
| Gene Symbol: ZBTB20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038993.1 | 6, 7 | 935 | |||
| GenBank mRNA | AK098660.1 | - | 6, 7 | 911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBTB20-AS1| Gene Symbol: ZBTB20-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038993.1 | 6, 7 | 935 | |||
| GenBank mRNA | AK098660.1 | - | 6, 7 | 911 | ||