siRNA Details
Gene Symbol: LINC00951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038887.1 | 2 | 2669 | |||
GenBank mRNA | BC132803.1 | - | 2 | 409 | ||
BC132805.1 | - | 2 | 409 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038887.1 | 2 | 2669 | |||
GenBank mRNA | BC132803.1 | - | 2 | 409 | ||
BC132805.1 | - | 2 | 409 |