siRNA Details
Gene Symbol: NFIA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104180.1 | 2 | 128 | |||
GenBank mRNA | HY006887.1 | - | 2 | 131 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL038993.1 | - | 2 | 124 | Chr. 1 - 61248999 - 61253512 [-] on Build GRCh38 |
