siRNA Details
| Gene Symbol: LOC101927120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110136.1 | 4 | 863 | |||
| NR_110137.1 | 3 | 743 | ||||
| GenBank mRNA | AK096987.1 | - | 4 | 863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927120| Gene Symbol: LOC101927120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110136.1 | 4 | 863 | |||
| NR_110137.1 | 3 | 743 | ||||
| GenBank mRNA | AK096987.1 | - | 4 | 863 | ||