siRNA Details
| Gene Symbol: DLGAP4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109939.1 | 3 | 539 | |||
| GenBank mRNA | BC039668.1 | - | 3 | 539 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830398.1 | - | 3 | 540 | Chr. 20 - 36508160 - 36573275 [-] on Build GRCh38 | |

DLGAP4-AS1
Genomic Map