siRNA Details
| Gene Symbol: LINC01592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039986.1 | 5 | 1203 | |||
| GenBank mRNA | AK055514.1 | - | 5 | 1203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01592| Gene Symbol: LINC01592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039986.1 | 5 | 1203 | |||
| GenBank mRNA | AK055514.1 | - | 5 | 1203 | ||