siRNA Details
| Gene Symbol: LOC101926897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120416.1 | 3 | 737 | |||
| GenBank mRNA | AK124674.1 | - | 3 | 738 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926897| Gene Symbol: LOC101926897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120416.1 | 3 | 737 | |||
| GenBank mRNA | AK124674.1 | - | 3 | 738 | ||