siRNA Details
| Gene Symbol: LOC101929315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125809.1 | 3 | 279 | |||
| GenBank mRNA | BX096372.1 | - | 3 | 239 | ||
| HY022654.1 | - | 2 | 401 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX107034.1 | - | 2 | 361 | Chr. 8 - 24511689 - 24514829 [-] on Build GRCh38 | |

LOC101929315
Genomic Map