siRNA Details
Gene Symbol: LOC101927189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125842.1 | 4 | 1074 | |||
GenBank mRNA | AK056584.1 | - | 4 | 875 | ||
BX640790.1 | - | 4 | 1013 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125842.1 | 4 | 1074 | |||
GenBank mRNA | AK056584.1 | - | 4 | 875 | ||
BX640790.1 | - | 4 | 1013 |