siRNA Details
| Gene Symbol: LOC101927189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125842.1 | 4 | 1338 | |||
| GenBank mRNA | AK056584.1 | - | 4 | 1139 | ||
| BX640790.1 | - | 4 | 1273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927189| Gene Symbol: LOC101927189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125842.1 | 4 | 1338 | |||
| GenBank mRNA | AK056584.1 | - | 4 | 1139 | ||
| BX640790.1 | - | 4 | 1273 | |||