siRNA Details
Gene Symbol: LOC105369187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135033.1 | 1 | 64 | |||
GenBank mRNA | BC043551.1 | - | 1 | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI831076.1 | - | 1 | 64 | Chr. 12 - 65544273 - 65557311 [-] on Build GRCh38 |
