siRNA Details
| Gene Symbol: LOC101927124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110045.1 | 4 | 589 | |||
| GenBank mRNA | BC041327.1 | - | 4 | 594 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI551679.1 | - | 4 | 593 | Chr. 14 - 31420756 - 31452479 [+] on Build GRCh38 | |

LOC101927124
Genomic Map