siRNA Details
| Gene Symbol: TRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040085.1 | 4 | 1240 | |||
| GenBank mRNA | AK125646.1 | - | 4 | 1202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRG-AS1| Gene Symbol: TRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040085.1 | 4 | 1240 | |||
| GenBank mRNA | AK125646.1 | - | 4 | 1202 | ||