siRNA Details
| Gene Symbol: DPP10-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132105.1 | 2 | 617 | |||
| GenBank mRNA | BC033566.1 | - | 2 | 617 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPP10-AS3| Gene Symbol: DPP10-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132105.1 | 2 | 617 | |||
| GenBank mRNA | BC033566.1 | - | 2 | 617 | ||