siRNA Details
| Gene Symbol: LINC00710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015413.1 | 6 | 731 | |||
| NR_024410.1 | 5 | 661 | ||||
| NR_024411.1 | 4 | 550 | ||||
| GenBank mRNA | BC033338.1 | - | 6 | 663 | ||
| BC106956.2 | - | 5 | 465 | |||
| BC106957.2 | - | 4 | 354 | |||
| BC113009.1 | - | 4 | 354 | |||

LINC00710
Genomic Map