siRNA Details
| Gene Symbol: LOC101927292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110048.1 | 2 | 286 | |||
| GenBank mRNA | BC034605.1 | - | 2 | 286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927292| Gene Symbol: LOC101927292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110048.1 | 2 | 286 | |||
| GenBank mRNA | BC034605.1 | - | 2 | 286 | ||