siRNA Details
| Gene Symbol: LOC100287015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040040.1 | 2 | 2360 | |||
| GenBank mRNA | AK025595.1 | - | 2 | 2358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100287015| Gene Symbol: LOC100287015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040040.1 | 2 | 2360 | |||
| GenBank mRNA | AK025595.1 | - | 2 | 2358 | ||