siRNA Details
Gene Symbol: LOC105373100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134637.1 | 2 | 1444 | |||
GenBank mRNA | BC050343.1 | - | 2 | 1414 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105373100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134637.1 | 2 | 1444 | |||
GenBank mRNA | BC050343.1 | - | 2 | 1414 |