siRNA Details
| Gene Symbol: LOC105370829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135679.1 | 2 | 216 | |||
| GenBank mRNA | BX103808.1 | - | 2 | 216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370829| Gene Symbol: LOC105370829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135679.1 | 2 | 216 | |||
| GenBank mRNA | BX103808.1 | - | 2 | 216 | ||