siRNA Details
| Gene Symbol: LINC01102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015399.1 | 2 | 257 | |||
| GenBank mRNA | AK055090.1 | - | 2 | 242 | ||
| AK126958.1 | - | 2 | 1337 | |||
| AL137513.1 | - | 2 | 226 | |||
| BC105699.1 | - | 2 | 151 | |||
| BC105700.1 | - | 2 | 151 | |||
| BC105701.1 | - | 2 | 151 | |||
| BC105702.1 | - | 2 | 151 | |||
| DA506317.1 | - | 2 | 258 | |||

LINC01102
Genomic Map