siRNA Details
| Gene Symbol: AGAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133447.1 | NP_597704.1 | 2 | 204 | ||
| GenBank mRNA | AK097165.1 | - | 2 | 204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGAP11| Gene Symbol: AGAP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133447.1 | NP_597704.1 | 2 | 204 | ||
| GenBank mRNA | AK097165.1 | - | 2 | 204 | ||