siRNA Details
| Gene Symbol: LOC101929586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120363.1 | 3 | 597 | |||
| GenBank mRNA | BC041477.1 | - | 3 | 597 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI914475.1 | - | 3 | 599 | Chr. 15 - 80556344 - 80562945 [-] on Build GRCh38 | |

LOC101929586
Genomic Map