siRNA Details
Gene Symbol: LOC101929312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110610.1 | 3 | 542 | |||
GenBank mRNA | BC043288.2 | - | 3 | 508 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI562295.1 | - | 3 | 512 | Chr. 20 - 7347467 - 7367626 [+] on Build GRCh38 |
