siRNA Details
| Gene Symbol: LOC100506675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120527.1 | 2 | 389 | |||
| GenBank mRNA | BX099517.1 | - | 2 | 353 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB024222.1 | - | 2 | 362 | Chr. 11 - 32040000 - 32041291 [-] on Build GRCh38 | |

LOC100506675
Genomic Map