siRNA Details
Gene Symbol: LINC00470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023925.1 | 3, 4 | 343 | |||
NR_110327.1 | 2, 3 | 287 | ||||
GenBank mRNA | AF295725.1 | - | 3, 4 | 343 | ||
AF295727.1 | - | 1, 2 | 46 | |||
AF295728.1 | - | 3, 4 | 163 | |||
AF295729.1 | - | 1, 2 | 46 |
