siRNA Details
Gene Symbol: MIR181A1HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF413001.1 | - | 2 | 2087 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR181A1HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF413001.1 | - | 2 | 2087 |