siRNA Details
| Gene Symbol: LOC284865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038460.1 | 3 | 799 | |||
| GenBank mRNA | AK092552.1 | - | 3 | 799 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284865| Gene Symbol: LOC284865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038460.1 | 3 | 799 | |||
| GenBank mRNA | AK092552.1 | - | 3 | 799 | ||