siRNA Details
Gene Symbol: LOC101929657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110306.1 | 1 | 2582 | |||
GenBank mRNA | BC036321.1 | - | 1 | 2586 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110306.1 | 1 | 2582 | |||
GenBank mRNA | BC036321.1 | - | 1 | 2586 |