siRNA Details
| Gene Symbol: LOC100129603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038729.1 | - | 4 | 472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129603| Gene Symbol: LOC100129603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038729.1 | - | 4 | 472 | ||