siRNA Details
Gene Symbol: LINC01305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038897.1 | 3 | 1952 | |||
GenBank mRNA | AL833646.1 | - | 3 | 1952 | ||
BC043589.1 | - | 2 | 565 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI913692.1 | - | 2 | 564 | Chr. 2 - 174328783 - 174329726 [+] on Build GRCh38 |
