siRNA Details
Gene Symbol: LOC100996455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073177.1 | 2 | 1131 | |||
GenBank mRNA | AK094364.1 | - | 2 | 1116 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073177.1 | 2 | 1131 | |||
GenBank mRNA | AK094364.1 | - | 2 | 1116 |