siRNA Details
| Gene Symbol: LOC101927412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125940.1 | 4 | 464 | |||
| NR_125941.1 | 3 | 416 | ||||
| NR_125942.1 | 3 | 343 | ||||
| GenBank mRNA | BU531772.1 | - | 3 | 346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927412| Gene Symbol: LOC101927412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125940.1 | 4 | 464 | |||
| NR_125941.1 | 3 | 416 | ||||
| NR_125942.1 | 3 | 343 | ||||
| GenBank mRNA | BU531772.1 | - | 3 | 346 | ||