siRNA Details
| Gene Symbol: LINC00691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026834.1 | 1 | 164 | |||
| GenBank mRNA | BC033355.1 | - | 1 | 155 | ||
| BC045572.1 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00691| Gene Symbol: LINC00691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026834.1 | 1 | 164 | |||
| GenBank mRNA | BC033355.1 | - | 1 | 155 | ||
| BC045572.1 | - | 1 | 140 | |||